Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042717.2 | NP_001036182.1 | 7 | 663 | ||
NM_033107.3 | NP_149098.2 | 9 | 900 | |||
GenBank mRNA | AF351613.1 | - | 7 | 664 | ||
AK095561.1 | - | 9 | 890 | |||
AK294325.1 | - | 7 | 613 | |||
BC004923.2 | - | 8 | 797 | |||
BC021573.1 | - | 8 | 797 | |||
BC107714.1 | - | 9 | 884 | 6714887 | ||
BC144162.1 | - | 7 | 610 | |||
CR933597.1 | - | 8 | 714 |