Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033109.4 | NP_149100.2 | 3, 4 | 386 | ||
XM_005264629.2 | XP_005264686.1 | 3, 4 | 316 | |||
XM_017005172.1 | XP_016860661.1 | 2, 3 | 266 | |||
GenBank mRNA | AJ458465.1 | - | 3, 4 | 317 | ||
AK307589.1 | - | 316 | ||||
AY027528.1 | - | 3, 4 | 313 | |||
BC053660.1 | - | 3, 4 | 300 | 6062060 | ||
DA998523.1 | - | 3, 4 | 352 | |||
DB461021.1 | - | 3, 4 | 386 |