Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HPS4 SIRNA Assays ›
Gene Symbol:
HPS4
Gene Name:
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases:
BLOC3S2, LE
Chromosome Location:
Chr.22: 26443613 - 26483863 on Build GRCh38
UniGene ID:
Hs.474436
Species:
Human
siRNA ID
HSS131782
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HPS4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022081.5
NP_071364.4
10
1398
NM_152841.2
NP_690054.1
8
1015
NR_073135.1
10
1416
NR_073136.1
9
1178
XM_006724353.3
XP_006724416.1
11
1443
XM_006724354.3
XP_006724417.1
11
1348
XM_006724360.3
XP_006724423.1
6
626
XM_011530485.1
XP_011528787.1
12
1521
XM_011530486.2
XP_011528788.1
12
1426
XM_011530487.2
XP_011528789.1
13
1631
XM_011530488.2
XP_011528790.1
12
1429
XM_011530489.2
XP_011528791.1
13
1532
XM_011530490.2
XP_011528792.1
11
1467
XM_011530491.2
XP_011528793.1
12
1521
XM_011530492.1
XP_011528794.1
12
1521
XM_011530493.2
XP_011528795.1
12
1521
XM_011530494.2
XP_011528796.1
10
817
XM_011530495.2
XP_011528797.1
4
323
XM_011530496.2
XP_011528798.1
5
158
XM_017029042.1
XP_016884531.1
10
1294
XM_017029043.1
XP_016884532.1
11
1499
XM_017029044.1
XP_016884533.1
10
1297
XM_017029045.1
XP_016884534.1
11
1467
XM_017029046.1
XP_016884535.1
10
1389
XM_017029047.1
XP_016884536.1
11
1467
XM_017029048.1
XP_016884537.1
10
1389
XM_017029049.1
XP_016884538.1
10
1294
XM_017029050.1
XP_016884539.1
10
1389
XM_017029051.1
XP_016884540.1
10
1294
XM_017029052.1
XP_016884541.1
11
1552
XM_017029053.1
XP_016884542.1
3
911
XM_017029054.1
XP_016884543.1
10
1498
XM_017029055.1
XP_016884544.1
10
1352
XM_017029056.1
XP_016884545.1
11
1565
XM_017029057.1
XP_016884546.1
11
1414
XM_017029058.1
XP_016884547.1
11
1448
XM_017029059.1
XP_016884548.1
11
1323
XM_017029060.1
XP_016884549.1
11
1322
XM_017029061.1
XP_016884550.1
11
1523
XM_017029062.1
XP_016884551.1
9
1298
XM_017029063.1
XP_016884552.1
11
1432
XM_017029064.1
XP_016884553.1
9
1181
GenBank mRNA
AB051454.1
-
9
3588
AK054717.1
-
7
1496
AK057648.1
-
11
1281
AK123691.1
-
5
1125
AK290211.1
-
10
1318
AL365512.1
-
8
919
AL713795.1
-
10
1347
AY043416.1
-
9
918
BC028625.1
-
10
1412
BC050285.1
-
10
1389
BC065030.1
-
9
1120
6148557
CR456392.1
-
9
819
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting
lysosome organization
blood coagulation
hemostasis
melanocyte differentiation
positive regulation of eye pigmentation
protein stabilization
positive regulation of protein targeting to mitochondrion
protein binding
protein homodimerization activity
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-pfczg:80/100.66.78.229:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline