Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HPS4 SIRNA Assays ›
Gene Symbol:
HPS4
Gene Name:
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases:
BLOC3S2, LE
Chromosome Location:
Chr.22: 26443613 - 26483863 on Build GRCh38
UniGene ID:
Hs.474436
Species:
Human
siRNA ID
HSS131783
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HPS4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022081.5
NP_071364.4
10
1436
NM_152841.2
NP_690054.1
8
1053
NR_073135.1
10
1454
NR_073136.1
9
1216
XM_006724353.3
XP_006724416.1
11
1481
XM_006724354.3
XP_006724417.1
11
1386
XM_006724360.3
XP_006724423.1
6
664
XM_011530485.1
XP_011528787.1
12
1559
XM_011530486.2
XP_011528788.1
12
1464
XM_011530487.2
XP_011528789.1
13
1669
XM_011530488.2
XP_011528790.1
12
1467
XM_011530489.2
XP_011528791.1
13
1570
XM_011530490.2
XP_011528792.1
11
1505
XM_011530491.2
XP_011528793.1
12
1559
XM_011530492.1
XP_011528794.1
12
1559
XM_011530493.2
XP_011528795.1
12
1559
XM_011530494.2
XP_011528796.1
10
855
XM_011530495.2
XP_011528797.1
4
361
XM_011530496.2
XP_011528798.1
5
196
XM_017029042.1
XP_016884531.1
10
1332
XM_017029043.1
XP_016884532.1
11
1537
XM_017029044.1
XP_016884533.1
10
1335
XM_017029045.1
XP_016884534.1
11
1505
XM_017029046.1
XP_016884535.1
10
1427
XM_017029047.1
XP_016884536.1
11
1505
XM_017029048.1
XP_016884537.1
10
1427
XM_017029049.1
XP_016884538.1
10
1332
XM_017029050.1
XP_016884539.1
10
1427
XM_017029051.1
XP_016884540.1
10
1332
XM_017029052.1
XP_016884541.1
11
1590
XM_017029053.1
XP_016884542.1
3
949
XM_017029054.1
XP_016884543.1
10
1536
XM_017029055.1
XP_016884544.1
10
1390
XM_017029056.1
XP_016884545.1
11
1603
XM_017029057.1
XP_016884546.1
11
1452
XM_017029058.1
XP_016884547.1
11
1486
XM_017029059.1
XP_016884548.1
11
1361
XM_017029060.1
XP_016884549.1
11
1360
XM_017029061.1
XP_016884550.1
11
1561
XM_017029062.1
XP_016884551.1
9
1336
XM_017029063.1
XP_016884552.1
11
1470
XM_017029064.1
XP_016884553.1
9
1219
GenBank mRNA
AB051454.1
-
9
3626
AK054717.1
-
7
1534
AK057648.1
-
11
1319
AK123691.1
-
5
1163
AK290211.1
-
10
1356
AL365512.1
-
8
957
AL713795.1
-
10
1385
AY043416.1
-
9
956
BC028625.1
-
10
1450
BC050285.1
-
10
1427
BC065030.1
-
9
1158
6148557
CR456392.1
-
9
857
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting
lysosome organization
blood coagulation
hemostasis
melanocyte differentiation
positive regulation of eye pigmentation
protein stabilization
positive regulation of protein targeting to mitochondrion
protein binding
protein homodimerization activity
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-nlx55:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline