Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171156.1 | NP_001164627.1 | 5 | 829 | ||
NM_001171157.1 | NP_001164628.1 | 5 | 1003 | |||
NM_001171158.1 | NP_001164629.1 | 5 | 859 | |||
NM_001171159.1 | NP_001164630.1 | 5 | 829 | |||
NM_033130.4 | NP_149121.2 | 5 | 1003 | |||
XM_011527439.1 | XP_011525741.1 | 5 | 687 | |||
GenBank mRNA | AF301007.1 | - | 5 | 898 | ||
AF310233.1 | - | 5 | 818 | |||
AF311905.1 | - | 5 | 1003 | |||
AK289900.1 | - | 5 | 789 | |||
AK290542.1 | - | 5 | 662 | |||
AK292025.1 | - | 5 | 906 | |||
AK294185.1 | - | 5 | 593 | |||
AK303514.1 | - | 5 | 698 | |||
AK308982.1 | - | 3 | 669 | |||
AY032685.1 | - | 5 | 763 | |||
AY358337.1 | - | 5 | 652 | |||
BC101725.1 | - | 5 | 868 | 8069231 | ||
BC143497.1 | - | 5 | 868 | 9052005 | ||
BC143500.1 | - | 5 | 694 | 9052008 |