Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167738.1 | NP_001161210.1 | 5, 6 | 1810 | ||
NM_020443.4 | NP_065176.3 | 7, 8 | 3141 | |||
XM_006711609.2 | XP_006711672.2 | 5, 6 | 1776 | |||
XM_006711610.2 | XP_006711673.2 | 5, 6 | 1776 | |||
XM_006711611.1 | XP_006711674.1 | 5, 6 | 1892 | |||
XM_011510097.1 | XP_011508399.1 | 7, 8 | 2820 | |||
XM_011510098.1 | XP_011508400.1 | 7, 8 | 2820 | |||
XM_011510099.1 | XP_011508401.1 | 7, 8 | 2820 | |||
XM_011510100.1 | XP_011508402.1 | 7, 8 | 2820 | |||
XM_011510101.1 | XP_011508403.1 | 7, 8 | 2820 | |||
XM_011510102.1 | XP_011508404.1 | 7, 8 | 2820 | |||
XM_017002751.1 | XP_016858240.1 | 7, 8 | 2820 | |||
XM_017002752.1 | XP_016858241.1 | 5, 6 | 1776 | |||
XM_017002753.1 | XP_016858242.1 | 7, 8 | 2820 | |||
XM_017002754.1 | XP_016858243.1 | 7, 8 | 2820 | |||
GenBank mRNA | AB032977.3 | - | 7, 8 | 2474 | ||
AB033039.1 | - | 2, 3 | 677 | |||
AJ488101.1 | - | 7, 8 | 2672 | |||
AK022631.1 | - | 4, 5 | 1567 | |||
AK056158.1 | - | 5, 6 | 1887 | |||
AY043013.1 | - | 7, 8 | 3141 | |||
BX648460.1 | - | 5, 6 | 1777 |