Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167738.1 | NP_001161210.1 | 13 | 2440 | ||
NM_020443.4 | NP_065176.3 | 15 | 3771 | |||
XM_006711609.2 | XP_006711672.2 | 13 | 2406 | |||
XM_006711610.2 | XP_006711673.2 | 12 | 2382 | |||
XM_006711611.1 | XP_006711674.1 | 13 | 2522 | |||
XM_011510097.1 | XP_011508399.1 | 15 | 3450 | |||
XM_011510098.1 | XP_011508400.1 | 14 | 3426 | |||
XM_011510099.1 | XP_011508401.1 | 14 | 3426 | |||
XM_011510100.1 | XP_011508402.1 | 14 | 3279 | |||
XM_011510101.1 | XP_011508403.1 | 13 | 3255 | |||
XM_011510102.1 | XP_011508404.1 | 13 | 3255 | |||
XM_017002751.1 | XP_016858240.1 | 14 | 3279 | |||
XM_017002752.1 | XP_016858241.1 | 12 | 2382 | |||
XM_017002753.1 | XP_016858242.1 | 14 | 3426 | |||
GenBank mRNA | AB032977.3 | - | 15 | 3104 | ||
AB033039.1 | - | 9 | 1283 | |||
AJ488101.1 | - | 15 | 3302 | |||
AK022631.1 | - | 11 | 2173 | |||
AK056158.1 | - | 13 | 2568 | |||
AY043013.1 | - | 15 | 3771 | |||
BC007523.1 | - | 2 | 62 | |||
BX648460.1 | - | 13 | 2407 | |||
CR936811.1 | - | 5 | 4164 |