Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193531.1 | NP_001180460.1 | 5, 6 | 563 | ||
NM_001294314.1 | NP_001281243.1 | 7, 8 | 780 | |||
NM_138341.2 | NP_612350.1 | 4, 5 | 499 | |||
NR_122119.1 | 4, 5 | 499 | ||||
XM_006719678.3 | XP_006719741.1 | 5, 6 | 465 | |||
XM_011538946.1 | XP_011537248.1 | 5, 6 | 417 | |||
XM_011538948.2 | XP_011537250.1 | 5, 6 | 417 | |||
XM_011538949.2 | XP_011537251.1 | 5, 6 | 368 | |||
XM_011538950.1 | XP_011537252.1 | 5, 6 | 417 | |||
XM_011538951.2 | XP_011537253.1 | 4, 5 | 336 | |||
XM_011538953.1 | XP_011537255.1 | 7, 8 | 753 | |||
XM_011538955.1 | XP_011537257.1 | 4, 5 | 360 | |||
XM_011538957.1 | XP_011537259.1 | 6, 7 | 674 | |||
XM_017020187.1 | XP_016875676.1 | 5, 6 | 417 | |||
XM_017020188.1 | XP_016875677.1 | 6, 7 | 637 | |||
XM_017020189.1 | XP_016875678.1 | 5, 6 | 525 | |||
XM_017020190.1 | XP_016875679.1 | 6, 7 | 582 | |||
XM_017020191.1 | XP_016875680.1 | 4, 5 | 446 | |||
XM_017020192.1 | XP_016875681.1 | 6, 7 | 2087 | |||
GenBank mRNA | AK074648.1 | - | 7, 8 | 692 | ||
AL519857.3 | - | 4, 5 | 415 | |||
AL833501.1 | - | 5, 6 | 484 | |||
BC048796.1 | - | 5, 6 | 455 | 5193616 | ||
BI828772.1 | - | 5, 6 | 563 |