Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138342.3 | NP_612351.2 | 19 | 2050 | ||
XM_006718937.1 | XP_006719000.1 | 19 | 1990 | |||
GenBank mRNA | AF173889.1 | - | 18 | 1677 | ||
AK074703.1 | - | 19 | 1935 | |||
AK074750.1 | - | 10 | 886 | |||
AK074855.1 | - | 19 | 1920 | |||
AK074947.1 | - | 19 | 1874 | |||
AY358497.1 | - | 19 | 1930 | |||
BC008326.1 | - | 15 | 1337 | |||
BC040641.1 | - | 19 | 2012 | 3849776 | ||
BX640934.1 | - | 20 | 4193 |