Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304437.1 | NP_001291366.1 | 3 | 429 | ||
NM_198920.2 | NP_944602.1 | 3 | 467 | |||
XM_011536238.2 | XP_011534540.1 | 3 | 462 | |||
XM_011536239.2 | XP_011534541.1 | 3 | 461 | |||
XM_011536240.1 | XP_011534542.1 | 3 | 461 | |||
XM_011536241.2 | XP_011534543.1 | 3 | 461 | |||
XM_011536242.1 | XP_011534544.1 | 3 | 465 | |||
XM_017011458.1 | XP_016866947.1 | 3 | 461 | |||
XM_017011459.1 | XP_016866948.1 | 3 | 461 | |||
XM_017011460.1 | XP_016866949.1 | 3 | 525 | |||
XM_017011461.1 | XP_016866950.1 | 3 | 461 | |||
GenBank mRNA | AB126062.1 | - | 3 | 452 | ||
AK298205.1 | - | 3 | 467 | |||
AL137544.1 | - | 2 | 214 | |||
BC036049.1 | - | 4 | 531 | |||
BC053645.1 | - | 3 | 457 | |||
BC057845.1 | - | 3 | 429 | |||
BC101512.1 | - | 3 | 377 | 8069018 | ||
BC101538.1 | - | 3 | 377 | 8069044 | ||
BC143617.1 | - | 3 | 377 |