Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130783.4 | NP_570139.3 | 4 | 335 | ||
XM_005253217.3 | XP_005253274.1 | 6 | 519 | |||
XM_006718372.3 | XP_006718435.1 | 6 | 606 | |||
XM_006718373.3 | XP_006718436.1 | 5 | 592 | |||
XM_011520459.2 | XP_011518761.1 | 5 | 482 | |||
GenBank mRNA | AK027715.1 | - | 3 | 144 | ||
AK075547.1 | - | 2 | 68 | |||
AY358087.1 | - | 3 | 258 | |||
BC019342.2 | - | 2 | 107 | 3357734 | ||
DA734054.1 | - | 5 | 385 |