Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284267.1 | NP_001271196.1 | 5, 6 | 383 | ||
NM_001284269.1 | NP_001271198.1 | 5, 6 | 541 | |||
NM_145231.3 | NP_660274.1 | 5, 6 | 530 | |||
GenBank mRNA | AK094740.1 | - | 5, 6 | 483 | ||
AL832321.1 | - | 5, 6 | 467 | |||
BC001787.1 | - | 5, 6 | 442 | 3353520 | ||
BP371056.1 | - | 5, 6 | 383 | |||
CF593330.1 | - | 5, 6 | 526 |