Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130896.2 | NP_570966.2 | 5 | 588 | ||
NM_181510.2 | NP_852611.2 | 5 | 588 | |||
XM_017028119.1 | XP_016883608.1 | 5 | 567 | |||
GenBank mRNA | AF492015.1 | - | 5 | 588 | ||
AF492016.1 | - | 5 | 588 | |||
AL591715.1 | - | 4 | 483 | |||
GQ472209.1 | - | 5 | 588 |