Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052848.1 | NP_443080.1 | 2 | 605 | ||
XM_005259377.4 | XP_005259434.1 | 2 | 968 | |||
XM_017027442.1 | XP_016882931.1 | 2 | 939 | |||
GenBank mRNA | AL833726.1 | - | 1 | 440 | ||
BC011577.1 | - | 2 | 605 | 4651484 |