Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM41A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080652.3 | NP_542383.1 | 5 | 784 | ||
XM_017007437.1 | XP_016862926.1 | 4 | 622 | |||
GenBank mRNA | AK290878.1 | - | 5 | 762 | ||
AK313954.1 | - | 5 | 720 | |||
AL832782.1 | - | 2 | 243 | |||
AY358547.1 | - | 5 | 768 | |||
BC008043.2 | - | 5 | 741 | 3838280 | ||
BC019884.1 | - | 5 | 731 | 4991416 | ||
BX647128.1 | - | 3 | 434 |