Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267774.1 | NP_001254703.1 | 5 | 626 | ||
NM_001267775.1 | NP_001254704.1 | 5 | 425 | |||
NM_001267776.1 | NP_001254705.1 | 4 | 389 | |||
NM_001267777.1 | NP_001254706.1 | 5 | 425 | |||
NM_174887.3 | NP_777547.1 | 5 | 507 | |||
GenBank mRNA | AF070643.1 | - | 1 | 558 | ||
AY224601.1 | - | 5 | 575 | |||
BC002640.2 | - | 5 | 487 | 3609552 | ||
BC038094.1 | - | 4 | 355 | 3907361 | ||
BE619475.1 | - | 5 | 361 | |||
BQ639919.1 | - | 5 | 375 |