Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033414.2 | NP_219482.1 | 2 | 776 | ||
GenBank mRNA | AY046059.1 | - | 2 | 787 | ||
BC008752.2 | - | 2 | 776 | 3345757 | ||
BC010545.1 | - | 2 | 765 | 3463070 |