Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039671.2 | NP_001034760.1 | 4, 5 | 522 | ||
NM_001039672.2 | NP_001034761.1 | 4, 5 | 540 | |||
NM_001039673.2 | NP_001034762.1 | 4, 5 | 531 | |||
NM_001145461.1 | NP_001138933.1 | 4, 5 | 516 | |||
NM_001145462.1 | NP_001138934.1 | 4, 5 | 492 | |||
NM_001145463.1 | NP_001138935.1 | 4, 5 | 531 | |||
XM_005259385.3 | XP_005259442.1 | 4, 5 | 753 | |||
XM_017027449.1 | XP_016882938.1 | 4, 5 | 541 | |||
XM_017027450.1 | XP_016882939.1 | 4, 5 | 539 | |||
XM_017027451.1 | XP_016882940.1 | 4, 5 | 537 | |||
GenBank mRNA | AF258578.3 | - | 3, 4 | 714 | ||
AK304479.1 | - | 2 | 878 | |||
AL833382.1 | - | 4, 5 | 488 | |||
AY358965.1 | - | 3, 4 | 659 | |||
BC007644.1 | - | 4, 5 | 479 | |||
BC014974.1 | - | 4, 5 | 485 | 4865479 | ||
BC091477.1 | - | 4, 5 | 514 | 30344388 | ||
BE877226.1 | - | 4, 5 | 468 | |||
BG765536.1 | - | 4, 5 | 473 | |||
BX379501.2 | - | 4, 5 | 458 | |||
CA487891.1 | - | 4, 5 | 474 | |||
CA488303.1 | - | 4, 5 | 469 | |||
CF127536.1 | - | 4, 5 | 486 |