Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DUOXA1 SIRNA Assays ›
Gene Symbol:
DUOXA1
Gene Name:
dual oxidase maturation factor 1
Gene Aliases:
NIP, NUMBIP, mol
Chromosome Location:
Chr.15: 45117366 - 45129938 on Build GRCh38
UniGene ID:
Hs.356664
Species:
Human
siRNA ID
HSS131995
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DUOXA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276264.1
NP_001263193.1
8
1127
NM_001276265.1
NP_001263194.1
4
649
NM_001276266.1
NP_001263195.1
5
784
NM_001276267.1
NP_001263196.1
8
1015
NM_001276268.1
NP_001263197.1
5
675
NM_144565.3
NP_653166.2
8
1157
XM_006720743.3
XP_006720806.1
8
1345
XM_006720745.1
XP_006720808.1
8
1320
XM_006720746.1
XP_006720809.1
8
1349
XM_006720747.3
XP_006720810.1
7
1257
XM_006720749.1
XP_006720812.1
8
1115
XM_006720751.3
XP_006720814.1
6
838
XM_006720752.3
XP_006720815.2
6
847
XM_006720754.1
XP_006720817.1
7
980
XM_006720755.1
XP_006720818.1
7
956
XM_011522181.2
XP_011520483.1
8
1373
XM_011522182.1
XP_011520484.1
7
1088
XM_011522183.2
XP_011520485.1
9
1203
XM_011522184.1
XP_011520486.1
6
804
GenBank mRNA
AK056896.1
-
5
675
AK292771.1
-
8
1127
BC020841.1
-
8
1015
4689593
BC029819.1
-
8
1155
5177619
DQ489735.1
-
5
761
EU927394.1
-
5
755
EU927395.1
-
4
620
EU927396.1
-
5
755
EU927397.1
-
4
620
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidase
oxidoreductase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
protein transport
hydrogen peroxide metabolic process
positive regulation of neuron differentiation
regulation of inflammatory response
positive regulation of reactive oxygen species metabolic process
regulation of thyroid hormone generation
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-wqf8l:80/100.66.76.116:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline