Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUOXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276264.1 | NP_001263193.1 | 6 | 653 | ||
NM_001276266.1 | NP_001263195.1 | 3 | 310 | |||
NM_144565.3 | NP_653166.2 | 6 | 683 | |||
XM_006720743.3 | XP_006720806.1 | 6 | 871 | |||
XM_006720745.1 | XP_006720808.1 | 6 | 846 | |||
XM_006720746.1 | XP_006720809.1 | 6 | 875 | |||
XM_006720747.3 | XP_006720810.1 | 5 | 783 | |||
XM_006720749.1 | XP_006720812.1 | 6 | 641 | |||
XM_006720751.3 | XP_006720814.1 | 4 | 364 | |||
XM_006720752.3 | XP_006720815.2 | 4 | 373 | |||
XM_011522181.2 | XP_011520483.1 | 6 | 899 | |||
XM_011522182.1 | XP_011520484.1 | 5 | 614 | |||
XM_011522183.2 | XP_011520485.1 | 7 | 729 | |||
XM_011522184.1 | XP_011520486.1 | 4 | 427 | |||
XM_011522185.1 | XP_011520487.1 | 4 | 427 | |||
XM_011522186.2 | XP_011520488.1 | 4 | 427 | |||
GenBank mRNA | AK292771.1 | - | 6 | 653 | ||
BC029819.1 | - | 6 | 681 | 5177619 | ||
DQ489735.1 | - | 3 | 287 | |||
EU927394.1 | - | 3 | 281 | |||
EU927396.1 | - | 3 | 281 |