Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARMIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138360.3 | NP_612369.3 | 32 | 3193 | ||
XM_011537300.2 | XP_011535602.1 | 31 | 2855 | |||
XM_017021751.1 | XP_016877240.1 | 28 | 2756 | |||
GenBank mRNA | AK074167.1 | - | 17 | 2566 | ||
AL834197.1 | - | 4 | 1390 |