Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRSAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005373.3 | NP_001005373.1 | 4 | 470 | ||
NM_001005374.3 | NP_001005374.1 | 3 | 175 | |||
NM_001190723.2 | NP_001177652.1 | 4 | 470 | |||
NM_138361.5 | NP_612370.3 | 3 | 729 | |||
XM_006717316.3 | XP_006717379.1 | 4 | 470 | |||
XM_017015283.1 | XP_016870772.1 | 4 | 1469 | |||
XM_017015284.1 | XP_016870773.1 | 4 | 470 | |||
GenBank mRNA | AK056203.1 | - | 4 | 452 | ||
AK091589.1 | - | 3 | 477 | |||
AK226106.1 | - | 4 | 456 | |||
AY358830.1 | - | 4 | 435 | |||
BC009239.2 | - | 3 | 175 | 2988664 | ||
DA165729.1 | - | 4 | 470 | |||
DB165274.1 | - | 4 | 470 |