Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316990.1 | NP_001303919.1 | 14 | 1966 | ||
NM_138363.2 | NP_612372.1 | 15 | 2072 | |||
NR_133644.1 | 15 | 2095 | ||||
XM_005257779.3 | XP_005257836.1 | 15 | 2048 | |||
XM_005257780.2 | XP_005257837.1 | 14 | 1691 | |||
XM_006722166.3 | XP_006722229.1 | 15 | 1830 | |||
XM_017025287.1 | XP_016880776.1 | 15 | 3051 | |||
XM_017025288.1 | XP_016880777.1 | 14 | 1724 | |||
GenBank mRNA | AK057326.1 | - | 15 | 2046 | ||
AK290587.1 | - | 15 | 1812 | |||
AK297409.1 | - | 14 | 1667 | |||
AL832822.1 | - | 15 | 1748 | |||
BC009518.1 | - | 15 | 1768 | 3633884 |