Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199640.1 | NP_001186569.1 | 4 | 322 | ||
NM_001314044.1 | NP_001300973.1 | 4 | 309 | |||
NM_001314045.1 | NP_001300974.1 | 4 | 317 | |||
NM_001314046.1 | NP_001300975.1 | 4 | 322 | |||
NM_001314047.1 | NP_001300976.1 | 4 | 309 | |||
NM_033439.3 | NP_254274.1 | 4 | 322 | |||
XM_011518061.2 | XP_011516363.1 | 3 | 257 | |||
XM_017015285.1 | XP_016870774.1 | 6 | 1068 | |||
GenBank mRNA | AB024518.1 | - | 3 | 261 | ||
AK303943.1 | - | 4 | 299 | |||
AK313414.1 | - | 4 | 301 | |||
AY905581.1 | - | 3 | 244 | |||
BC047085.1 | - | 4 | 299 | 5314040 | ||
BQ878541.1 | - | 3 | 283 | |||
CR407619.1 | - | 3 | 244 | |||
DA439244.1 | - | 4 | 317 | |||
DA713547.1 | - | 4 | 309 | |||
DA719342.1 | - | 4 | 307 | |||
DB199599.1 | - | 4 | 331 | |||
DB266668.1 | - | 4 | 301 |