Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199640.1 | NP_001186569.1 | 6 | 485 | ||
NM_001199641.1 | NP_001186570.1 | 4 | 233 | |||
NM_001314044.1 | NP_001300973.1 | 7 | 598 | |||
NM_001314045.1 | NP_001300974.1 | 7 | 606 | |||
NM_001314046.1 | NP_001300975.1 | 7 | 593 | |||
NM_001314047.1 | NP_001300976.1 | 7 | 580 | |||
NM_001314048.1 | NP_001300977.1 | 6 | 485 | |||
NM_033439.3 | NP_254274.1 | 7 | 611 | |||
XM_011518061.2 | XP_011516363.1 | 6 | 546 | |||
XM_017015285.1 | XP_016870774.1 | 9 | 1339 | |||
GenBank mRNA | AB024518.1 | - | 6 | 550 | ||
AK295908.1 | - | 4 | 214 | |||
AK303943.1 | - | 6 | 462 | |||
AK313414.1 | - | 7 | 590 | |||
AY905581.1 | - | 6 | 533 | |||
BC047085.1 | - | 7 | 588 | 5314040 | ||
BQ878541.1 | - | 6 | 572 | |||
CR407619.1 | - | 6 | 533 | |||
DB266668.1 | - | 7 | 572 | |||
HQ641439.1 | - | 5 | 422 |