Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138961.2 | NP_620411.2 | 4, 5 | 784 | ||
XM_011543060.2 | XP_011541362.1 | 3, 4 | 547 | |||
GenBank mRNA | AF361746.1 | - | 4, 5 | 749 | ||
AK075396.1 | - | 4, 5 | 747 | |||
AK091023.1 | - | 4, 5 | 1667 | |||
AK092429.1 | - | 2, 3 | 1156 | |||
AK314710.1 | - | 4, 5 | 747 | |||
AY189281.1 | - | 1, 2 | 1330 | |||
AY358382.1 | - | 4, 5 | 736 | |||
BC016868.1 | - | 4, 5 | 618 | 3850665 |