Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052853.3 | NP_443085.2 | 1, 2 | 1102 | ||
XM_006716170.3 | XP_006716233.1 | 1, 2 | 1327 | |||
XM_011516675.2 | XP_011514977.1 | 1, 2 | 1327 | |||
GenBank mRNA | AY302592.1 | - | 1, 2 | 1102 | ||
BC014107.2 | - | 1, 2 | 1099 | 4138734 |