Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198963.1 | NP_945314.1 | 6 | 1568 | ||
XM_011533154.2 | XP_011531456.1 | 6 | 1572 | |||
XM_011533155.2 | XP_011531457.1 | 6 | 1570 | |||
XM_011533156.2 | XP_011531458.1 | 6 | 1572 | |||
XM_011533157.1 | XP_011531459.1 | 6 | 1572 | |||
GenBank mRNA | AB209249.1 | - | 1 | 35 | ||
AF283512.1 | - | 2 | 259 | |||
AK057423.1 | - | 5 | 1368 | |||
AK296739.1 | - | 6 | 1607 | |||
AK307514.1 | - | 6 | 1597 | |||
BC039270.1 | - | 5 | 1375 | |||
BC060778.1 | - | 6 | 1568 | |||
BC065278.1 | - | 1 | 63 | |||
BC131534.1 | - | 6 | 1540 | 40082400 | ||
BQ719095.1 | - | 2 | 431 | |||
BU183594.1 | - | 1 | 62 |