Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175736.4 | NP_783863.4 | 7, 8 | 942 | ||
NM_198900.2 | NP_944489.2 | 6, 7 | 789 | |||
XM_005269218.2 | XP_005269275.1 | 7, 8 | 921 | |||
XM_011538968.2 | XP_011537270.1 | 7, 8 | 1411 | |||
XM_011538969.2 | XP_011537271.1 | 7, 8 | 1410 | |||
XM_011538970.2 | XP_011537272.1 | 7, 8 | 1413 | |||
XM_011538971.2 | XP_011537273.1 | 7, 8 | 986 | |||
XM_011538972.2 | XP_011537274.1 | 6, 7 | 1256 | |||
XM_011538973.2 | XP_011537275.2 | 3, 4 | 401 | |||
XM_011538974.2 | XP_011537276.1 | 8, 9 | 743 | |||
GenBank mRNA | AB095934.2 | - | 7, 8 | 942 | ||
AK128195.1 | - | 6, 7 | 769 | |||
BC159100.1 | - | 7, 8 | 942 | 8860524 | ||
BQ707902.1 | - | 7, 8 | 603 |