Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139159.4 | NP_631898.3 | 4 | 480 | ||
XM_005259673.2 | XP_005259730.1 | 5 | 589 | |||
XM_011528405.1 | XP_011526707.1 | 6 | 641 | |||
XM_011528406.1 | XP_011526708.1 | 7 | 751 | |||
XM_011528408.1 | XP_011526710.1 | 4 | 426 | |||
XM_011528410.1 | XP_011526712.1 | 5 | 641 | |||
XM_017027453.1 | XP_016882942.1 | 6 | 1231 | |||
XM_017027454.1 | XP_016882943.1 | 7 | 1341 | |||
GenBank mRNA | AF452102.2 | - | 4 | 500 | ||
AF542510.2 | - | 4 | 459 | |||
AK122654.1 | - | 4 | 483 | |||
AK131100.1 | - | 1 | 49 | |||
AY172660.1 | - | 1 | 145 | |||
AY374518.1 | - | 1 | 135 | |||
BC037948.2 | - | 4 | 417 | 4577856 | ||
CB960096.1 | - | 5 | 605 | |||
DB105598.1 | - | 4 | 480 | |||
DQ417928.1 | - | 2 | 222 |