Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139159.4 | NP_631898.3 | 5 | 666 | ||
XM_005259673.2 | XP_005259730.1 | 6 | 775 | |||
XM_011528405.1 | XP_011526707.1 | 7 | 827 | |||
XM_011528406.1 | XP_011526708.1 | 8 | 937 | |||
XM_011528408.1 | XP_011526710.1 | 5 | 612 | |||
XM_011528410.1 | XP_011526712.1 | 6 | 827 | |||
XM_017027453.1 | XP_016882942.1 | 7 | 1417 | |||
XM_017027454.1 | XP_016882943.1 | 8 | 1527 | |||
GenBank mRNA | AF452102.2 | - | 5 | 686 | ||
AF542510.2 | - | 5 | 645 | |||
AK122654.1 | - | 5 | 669 | |||
AK131100.1 | - | 2 | 235 | |||
AY172660.1 | - | 2 | 331 | |||
AY374518.1 | - | 2 | 321 | |||
BC037948.2 | - | 5 | 603 | 4577856 | ||
DQ417928.1 | - | 3 | 408 |