Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130726.2 | NP_001124198.1 | 3, 4 | 366 | ||
NM_173463.4 | NP_775734.2 | 4, 5 | 467 | |||
XM_005248210.3 | XP_005248267.1 | 3, 4 | 395 | |||
XM_011513906.2 | XP_011512208.1 | 5, 6 | 477 | |||
XM_011513907.2 | XP_011512209.1 | 4, 5 | 350 | |||
XM_011513908.1 | XP_011512210.1 | 3, 4 | 272 | |||
XM_017008827.1 | XP_016864316.1 | 5, 6 | 477 | |||
XM_017008828.1 | XP_016864317.1 | 4, 5 | 401 | |||
GenBank mRNA | AK125353.1 | - | 3, 4 | 275 | ||
AK302905.1 | - | 4, 5 | 411 | |||
AL834257.2 | - | 3, 4 | 340 | |||
BC057761.1 | - | 3, 4 | 342 | |||
BC067735.1 | - | 3, 4 | 342 | |||
DC422585.1 | - | 3, 4 | 358 | |||
DR423861.1 | - | 3, 4 | 281 |