Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130726.2 | NP_001124198.1 | 9 | 1014 | ||
NM_173463.4 | NP_775734.2 | 10 | 1115 | |||
XM_005248210.3 | XP_005248267.1 | 9 | 1043 | |||
XM_011513906.2 | XP_011512208.1 | 11 | 1125 | |||
XM_011513907.2 | XP_011512209.1 | 10 | 998 | |||
XM_011513908.1 | XP_011512210.1 | 9 | 920 | |||
XM_017008827.1 | XP_016864316.1 | 11 | 1125 | |||
XM_017008828.1 | XP_016864317.1 | 10 | 1049 | |||
GenBank mRNA | AK125353.1 | - | 9 | 923 | ||
AK296454.1 | - | 6 | 672 | |||
AK302905.1 | - | 10 | 1059 | |||
AL834257.2 | - | 9 | 988 |