Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142301.1 | NP_001135773.1 | 7 | 630 | ||
NM_153704.5 | NP_714915.3 | 6 | 699 | |||
NR_024522.1 | 6 | 699 | ||||
XM_006716686.3 | XP_006716749.1 | 5 | 537 | |||
GenBank mRNA | AK092244.1 | - | 7 | 587 | ||
AK094935.1 | - | 6 | 623 | |||
AK308071.1 | - | 7 | 589 | |||
BC032835.2 | - | 6 | 617 | 4825770 | ||
BC054338.1 | - | 6 | 626 | |||
BX648768.1 | - | 6 | 643 | |||
KU178827.1 | - | 6 | 598 |