Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164586.1 | NP_001158058.1 | 19 | 10078 | ||
XM_005245580.3 | XP_005245637.1 | 19 | 10048 | |||
XM_006711618.3 | XP_006711681.1 | 19 | 10123 | |||
XM_011510120.2 | XP_011508422.1 | 19 | 10170 | |||
XM_011510121.2 | XP_011508423.1 | 19 | 10155 | |||
XM_017002787.1 | XP_016858276.1 | 18 | 9850 | |||
GenBank mRNA | AL137493.1 | - | 8 | 2052 | ||
AY245430.2 | - | 8 | 2200 | |||
BC033185.1 | - | 21 | 2657 | |||
BC131529.1 | - | 8 | 1781 | 40036345 |