Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033200.2 | NP_149977.2 | 9, 10 | 1279 | ||
XM_005261955.3 | XP_005262012.1 | 9, 10 | 1361 | |||
XM_006724426.3 | XP_006724489.1 | 8, 9 | 1725 | |||
XM_006724427.3 | XP_006724490.1 | 9, 10 | 1276 | |||
GenBank mRNA | BC002942.2 | - | 9, 10 | 1253 | 3954961 | |
BC014652.1 | - | 9, 10 | 1342 | 4562676 | ||
BC021143.1 | - | 9, 10 | 1253 | 4872642 | ||
CR456605.1 | - | 9, 10 | 1263 |