Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033026.1 | NP_001028198.1 | 2, 3 | 656 | ||
NM_033420.3 | NP_219488.1 | 2, 3 | 656 | |||
XM_005259676.2 | XP_005259733.1 | 2, 3 | 683 | |||
GenBank mRNA | AK095756.1 | - | 2, 3 | 656 | ||
BC008957.2 | - | 2, 3 | 629 | 2964663 | ||
BQ895919.1 | - | 2, 3 | 427 | |||
DQ005958.1 | - | 2, 3 | 638 |