Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN2AIPNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080656.2 | NP_542387.1 | 1 | 148 | ||
XM_011543709.2 | XP_011542011.1 | 2 | 293 | |||
GenBank mRNA | BC008293.1 | - | 1 | 139 | 3537789 | |
BC018086.1 | - | 1 | 120 | 4793897 | ||
DB480185.1 | - | 1 | 148 |