Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052855.3 | NP_443087.1 | 4 | 1057 | ||
XM_017025296.1 | XP_016880785.1 | 4 | 804 | |||
GenBank mRNA | AK314010.1 | - | 4 | 804 | ||
BC005853.2 | - | 2 | 86 | |||
BC012978.2 | - | 4 | 1049 | 3834331 | ||
CR627413.1 | - | 4 | 829 | |||
EU164539.1 | - | 4 | 1049 |