Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135825.1 | NP_001129297.1 | 4 | 427 | ||
NM_152267.3 | NP_689480.2 | 6 | 595 | |||
NR_024209.1 | 6 | 580 | ||||
NR_024210.1 | 6 | 762 | ||||
NR_024211.1 | 5 | 482 | ||||
NR_024212.1 | 5 | 371 | ||||
GenBank mRNA | AK027035.1 | - | 5 | 294 | ||
AK095947.1 | - | 4 | 336 | |||
AK291236.1 | - | 6 | 504 | |||
AK297414.1 | - | 5 | 452 | |||
BC009504.2 | - | 6 | 544 | 2964524 | ||
BC012817.1 | - | 6 | 553 | 2964524 | ||
BC026040.1 | - | 6 | 487 | 4866294 | ||
BC033166.1 | - | 6 | 530 | 4549320 | ||
BC035684.1 | - | 6 | 554 | 5750180 | ||
BX648019.1 | - | 6 | 504 | |||
CR456349.1 | - | 6 | 483 | |||
DA498404.1 | - | 4 | 400 | |||
DQ296559.1 | - | 6 | 536 | |||
DQ296561.1 | - | 6 | 536 | |||
DQ296562.1 | - | 4 | 368 | |||
DQ296564.1 | - | 6 | 704 | |||
DQ296565.1 | - | 6 | 536 |