Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCED1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281429.1 | NP_001268358.1 | 3 | 1366 | ||
NM_138371.2 | NP_612380.1 | 4 | 1834 | |||
XM_005269224.4 | XP_005269281.1 | 4 | 1531 | |||
XM_011538978.2 | XP_011537280.1 | 5 | 1699 | |||
XM_017020207.1 | XP_016875696.1 | 6 | 1756 | |||
XM_017020208.1 | XP_016875697.1 | 7 | 1833 | |||
XM_017020209.1 | XP_016875698.1 | 5 | 1588 | |||
XM_017020210.1 | XP_016875699.1 | 6 | 3505 | |||
XM_017020211.1 | XP_016875700.1 | 3 | 1785 | |||
XM_017020212.1 | XP_016875701.1 | 4 | 1528 | |||
XM_017020213.1 | XP_016875702.1 | 7 | 1901 | |||
XM_017020214.1 | XP_016875703.1 | 8 | 2017 | |||
XM_017020215.1 | XP_016875704.1 | 7 | 1872 | |||
XM_017020216.1 | XP_016875705.1 | 3 | 3076 | |||
GenBank mRNA | BC008360.1 | - | 2 | 1581 | 3610443 | |
BC016154.1 | - | 3 | 788 | 4040497 | ||
BC072670.1 | - | 4 | 1832 |