Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS19BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194326.3 | NP_919307.1 | 3 | 325 | ||
NR_130151.1 | 4 | 416 | ||||
GenBank mRNA | AK025423.1 | - | 3 | 218 | ||
AK055865.1 | - | 1 | 2203 | |||
BC037573.1 | - | 3 | 335 | 5399759 | ||
BC047711.1 | - | 3 | 217 | 6064917 | ||
CB142920.1 | - | 4 | 322 | |||
CR456443.1 | - | 3 | 255 |