Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLFN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104587.1 | NP_001098057.1 | 7 | 2814 | ||
NM_001104588.1 | NP_001098058.1 | 7 | 2732 | |||
NM_001104589.1 | NP_001098059.1 | 6 | 2654 | |||
NM_001104590.1 | NP_001098060.1 | 7 | 2644 | |||
NM_152270.3 | NP_689483.3 | 5 | 2537 | |||
XM_005258068.3 | XP_005258125.1 | 5 | 2642 | |||
XM_006722171.3 | XP_006722234.1 | 7 | 2625 | |||
XM_011525465.2 | XP_011523767.1 | 6 | 2847 | |||
XM_017025299.1 | XP_016880788.1 | 7 | 2965 | |||
GenBank mRNA | AK074184.1 | - | 5 | 2401 | ||
AK092241.1 | - | 7 | 2567 | |||
AL512731.1 | - | 1 | 67 | |||
AL831964.2 | - | 5 | 2437 | |||
BC052586.1 | - | 5 | 2443 | 6258140 |