Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMDT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033318.4 | NP_201575.3 | 1 | 154 | ||
XM_011530509.2 | XP_011528811.1 | 1 | 158 | |||
GenBank mRNA | AK312142.1 | - | 1 | 145 | ||
AL449243.1 | - | 1 | 137 | |||
BC024237.2 | - | 1 | 150 | 4798793 | ||
BI754670.1 | - | 1 | 154 | |||
CR456453.1 | - | 1 | 94 |