Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080658.1 | NP_542389.1 | 6 | 778 | ||
XM_017018549.1 | XP_016874038.1 | 7 | 969 | |||
XM_017018550.1 | XP_016874039.1 | 7 | 1003 | |||
XM_017018551.1 | XP_016874040.1 | 7 | 924 | |||
GenBank mRNA | AF359506.1 | - | 5 | 697 | ||
AY032593.1 | - | 758 | ||||
AY040761.1 | - | 6 | 758 | |||
AY169233.1 | - | 6 | 778 | |||
BC008689.1 | - | 6 | 778 | 3853707 |