Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031733.2 | NP_001026903.2 | 4 | 1098 | ||
NM_001286694.1 | NP_001273623.1 | 4 | 1208 | |||
NM_001286695.1 | NP_001273624.1 | 3 | 1067 | |||
NM_033429.2 | NP_219501.2 | 5 | 1239 | |||
NR_104583.1 | 2 | 878 | ||||
GenBank mRNA | AK092628.1 | - | 4 | 320 | ||
AK296804.1 | - | 5 | 461 | |||
BC009516.2 | - | 4 | 415 | |||
BX640702.1 | - | 3 | 256 | |||
BX640817.1 | - | 2 | 879 | |||
DB204573.1 | - | 5 | 463 |