Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174889.4 | NP_777549.1 | 2, 3 | 339 | ||
GenBank mRNA | AB183433.1 | - | 2, 3 | 212 | ||
AF087990.1 | - | 2, 3 | 308 | |||
AK026051.1 | - | 2, 3 | 339 | |||
AK291296.1 | - | 2, 3 | 302 | |||
BC001753.1 | - | 1 | 261 | 3354845 | ||
BC033965.1 | - | 2, 3 | 255 | 5288040 | ||
KU178834.1 | - | 2, 3 | 212 | |||
KU178836.1 | - | 2, 3 | 212 |