Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183376.2 | NP_899232.2 | 5 | 995 | ||
GenBank mRNA | AK093364.1 | - | 5 | 575 | ||
AK290221.1 | - | 5 | 995 | |||
AK296448.1 | - | 2 | 325 | |||
AK316280.1 | - | 2 | 319 | |||
BC028704.2 | - | 5 | 985 | 4838787 | ||
BC070100.1 | - | 5 | 975 | 30345612 |