Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3HAV1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080660.3 | NP_542391.2 | 2 | 405 | ||
XM_006716176.3 | XP_006716239.1 | 2 | 848 | |||
XM_011516688.2 | XP_011514990.1 | 2 | 846 | |||
XM_011516689.2 | XP_011514991.1 | 2 | 750 | |||
GenBank mRNA | BC008842.2 | - | 2 | 405 | 4136104 |