Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033547.3 | NP_291025.3 | 4 | 465 | ||
XM_011545352.2 | XP_011543654.1 | 5 | 634 | |||
XM_011545353.2 | XP_011543655.1 | 4 | 643 | |||
XM_017018560.1 | XP_016874049.1 | 4 | 498 | |||
GenBank mRNA | AK000455.1 | - | 5 | 540 | ||
AK290866.1 | - | 4 | 461 | |||
AL834171.1 | - | 4 | 408 | |||
BC009995.2 | - | 4 | 460 | 4129693 |